The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030288.2 | NP_001025459.1 | 1 - 2 | 133 | 80 |
GenBank mRNA | DA951208.1 | - | 1 - 2 | 133 | 80 |
X60502.1 | - | 1 - 2 | 104 | 80 |