Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLMAP
Entrez Gene ID
7871
Gene Name
sarcolemma associated protein
Gene Aliases
SLAP
Chromosome Location
Chr.3: 57756221 - 57930013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007159.4 NP_009090.2 10 - 11 1536 61
XM_005265465.3 XP_005265522.1 11 - 12 2372 61
XM_005265466.3 XP_005265523.1 11 - 12 2366 61
XM_006713326.2 XP_006713389.1 11 - 12 2366 61
XM_011534104.1 XP_011532406.1 11 - 12 2372 61
XM_017007151.1 XP_016862640.1 11 - 12 2372 61
XM_017007152.1 XP_016862641.1 11 - 12 2366 61
XM_017007164.1 XP_016862653.1 11 - 12 2366 61
GenBank mRNA AB046821.1 - 8 - 9 777 61

View Full Product Details