Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLMAP
Entrez Gene ID
7871
Gene Name
sarcolemma associated protein
Gene Aliases
SLAP
Chromosome Location
Chr.3: 57756221 - 57930013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304420.2 NP_001291349.1 13 - 14 1703 106
NM_001304421.2 NP_001291350.1 11 - 12 1589 106
NM_001304422.2 NP_001291351.1 1 - 2 185 106
NM_001304423.2 NP_001291352.1 1 - 2 185 106
NM_001311178.1 NP_001298107.1 1 - 2 185 106
NM_001311179.1 NP_001298108.1 1 - 2 185 106
NM_007159.4 NP_009090.2 12 - 13 1652 106
XM_005265456.3 XP_005265513.1 15 - 16 2590 106
XM_005265457.3 XP_005265514.1 15 - 16 2584 106
XM_005265458.3 XP_005265515.1 15 - 16 2590 106
XM_005265459.3 XP_005265516.1 14 - 15 2539 106
XM_005265460.3 XP_005265517.1 14 - 15 2539 106
XM_005265461.3 XP_005265518.1 14 - 15 2533 106
XM_005265462.3 XP_005265519.1 14 - 15 2527 106
XM_005265464.3 XP_005265521.1 14 - 15 2521 106
XM_005265465.3 XP_005265522.1 13 - 14 2488 106
XM_005265466.3 XP_005265523.1 13 - 14 2482 106
XM_005265467.3 XP_005265524.1 13 - 14 2476 106
XM_005265468.3 XP_005265525.1 15 - 16 2590 106
XM_005265470.3 XP_005265527.1 12 - 13 2425 106
XM_005265471.3 XP_005265528.1 12 - 13 2419 106
XM_005265472.3 XP_005265529.1 13 - 14 2476 106
XM_006713326.2 XP_006713389.1 13 - 14 2482 106
XM_011534103.1 XP_011532405.1 13 - 14 2476 106
XM_011534104.1 XP_011532406.1 13 - 14 2488 106
XM_011534105.1 XP_011532407.1 12 - 13 2425 106
XM_017007146.1 XP_016862635.1 15 - 16 2584 106
XM_017007147.1 XP_016862636.1 14 - 15 2533 106
XM_017007148.1 XP_016862637.1 14 - 15 2539 106
XM_017007149.1 XP_016862638.1 14 - 15 2533 106
XM_017007150.1 XP_016862639.1 14 - 15 2533 106
XM_017007151.1 XP_016862640.1 13 - 14 2488 106
XM_017007152.1 XP_016862641.1 13 - 14 2482 106
XM_017007153.1 XP_016862642.1 13 - 14 2470 106
XM_017007154.1 XP_016862643.1 13 - 14 2470 106
XM_017007155.1 XP_016862644.1 15 - 16 2584 106
XM_017007156.1 XP_016862645.1 15 - 16 2590 106
XM_017007157.1 XP_016862646.1 15 - 16 2584 106
XM_017007158.1 XP_016862647.1 12 - 13 2425 106
XM_017007159.1 XP_016862648.1 14 - 15 2539 106
XM_017007160.1 XP_016862649.1 12 - 13 2419 106
XM_017007161.1 XP_016862650.1 14 - 15 2533 106
XM_017007162.1 XP_016862651.1 14 - 15 2539 106
XM_017007163.1 XP_016862652.1 14 - 15 2533 106
XM_017007164.1 XP_016862653.1 13 - 14 2482 106
XM_017007165.1 XP_016862654.1 13 - 14 2470 106
XM_017007166.1 XP_016862655.1 13 - 14 2470 106
XM_017007167.1 XP_016862656.1 12 - 13 2425 106
XM_017007168.1 XP_016862657.1 12 - 13 2419 106
XM_017007169.1 XP_016862658.1 15 - 16 2590 106
GenBank mRNA AB046821.1 - 10 - 11 893 106
AF304450.1 - 1 - 2 58 106
AK124200.1 - 13 - 14 1393 106
AK300106.1 - 1 - 2 108 106
AK304493.1 - 1 - 2 112 106
BC114627.1 - 11 - 12 1287 106
BC115701.1 - 13 - 14 1401 106
DB100357.1 - 1 - 2 189 106

View Full Product Details