Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLMAP
Entrez Gene ID
7871
Gene Name
sarcolemma associated protein
Gene Aliases
SLAP
Chromosome Location
Chr.3: 57756221 - 57930013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304420.2 NP_001291349.1 16 - 17 1970 126
NM_001304421.2 NP_001291350.1 14 - 15 1856 126
NM_001304422.2 NP_001291351.1 4 - 5 452 126
NM_001311178.1 NP_001298107.1 4 - 5 452 126
NM_007159.4 NP_009090.2 15 - 16 1919 126
XM_005265456.3 XP_005265513.1 18 - 19 2857 126
XM_005265457.3 XP_005265514.1 18 - 19 2851 126
XM_005265458.3 XP_005265515.1 18 - 19 2857 126
XM_005265459.3 XP_005265516.1 17 - 18 2806 126
XM_005265460.3 XP_005265517.1 17 - 18 2806 126
XM_005265461.3 XP_005265518.1 17 - 18 2800 126
XM_005265462.3 XP_005265519.1 17 - 18 2794 126
XM_005265464.3 XP_005265521.1 17 - 18 2788 126
XM_005265465.3 XP_005265522.1 16 - 17 2755 126
XM_005265466.3 XP_005265523.1 16 - 17 2749 126
XM_005265467.3 XP_005265524.1 16 - 17 2743 126
XM_005265470.3 XP_005265527.1 15 - 16 2692 126
XM_005265471.3 XP_005265528.1 15 - 16 2686 126
XM_011534103.1 XP_011532405.1 16 - 17 2743 126
XM_011534106.1 XP_011532408.1 4 - 5 327 126
XM_017007146.1 XP_016862635.1 18 - 19 2851 126
XM_017007147.1 XP_016862636.1 17 - 18 2800 126
XM_017007148.1 XP_016862637.1 17 - 18 2806 126
XM_017007149.1 XP_016862638.1 17 - 18 2800 126
XM_017007150.1 XP_016862639.1 17 - 18 2800 126
XM_017007151.1 XP_016862640.1 16 - 17 2755 126
XM_017007152.1 XP_016862641.1 16 - 17 2749 126
XM_017007153.1 XP_016862642.1 16 - 17 2737 126
XM_017007154.1 XP_016862643.1 16 - 17 2737 126
XM_017007158.1 XP_016862647.1 15 - 16 2692 126
XM_017007160.1 XP_016862649.1 15 - 16 2686 126
XM_017007170.1 XP_016862659.1 4 - 5 327 126
GenBank mRNA AB046821.1 - 13 - 14 1160 126
AF100750.1 - 4 - 5 396 126
AK304493.1 - 4 - 5 379 126
AK316436.1 - 4 - 5 320 126
BC114627.1 - 14 - 15 1554 126
BC115701.1 - 16 - 17 1668 126
DB100357.1 - 4 - 5 456 126

View Full Product Details