The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018060.3 | NP_060530.3 | 10 - 11 | 1427 | 107 |
GenBank mRNA | AK001188.1 | - | 8 - 9 | 902 | 107 |
AK001726.1 | - | 4 - 5 | 416 | 107 | |
AK291432.1 | - | 10 - 11 | 1236 | 107 | |
AY267462.1 | - | 10 - 11 | 1438 | 107 | |
BC010218.1 | - | 6 - 7 | 607 | 107 | |
BC040376.1 | - | 10 - 11 | 1160 | 107 | |
BC047880.1 | - | 9 - 10 | 954 | 107 | |
BC137438.1 | - | 10 - 11 | 1397 | 107 | |
D28500.1 | - | 10 - 11 | 1268 | 107 |