The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128225.2 | NP_001121697.1 | 6 - 7 | 916 | 58 |
NM_152264.4 | NP_689477.2 | 6 - 7 | 937 | 58 | |
NR_134854.1 | 6 - 7 | 1006 | 58 | ||
XM_006718381.3 | XP_006718444.1 | 6 - 7 | 821 | 58 | |
XM_006718383.3 | XP_006718446.1 | 5 - 6 | 713 | 58 | |
XM_006718384.2 | XP_006718447.1 | 6 - 7 | 821 | 58 | |
XM_006718385.2 | XP_006718448.1 | 5 - 6 | 713 | 58 | |
XM_011520466.1 | XP_011518768.1 | 6 - 7 | 821 | 58 | |
XM_011520467.1 | XP_011518769.1 | 6 - 7 | 1150 | 58 | |
XM_011520468.2 | XP_011518770.1 | 6 - 7 | 1243 | 58 | |
XM_011520469.1 | XP_011518771.1 | 5 - 6 | 713 | 58 | |
XM_011520470.1 | XP_011518772.1 | 5 - 6 | 687 | 58 | |
XM_017018540.1 | XP_016874029.1 | 6 - 7 | 795 | 58 | |
XM_017018541.1 | XP_016874030.1 | 5 - 6 | 687 | 58 | |
XM_017018542.1 | XP_016874031.1 | 6 - 7 | 795 | 58 | |
GenBank mRNA | AK098651.1 | - | 6 - 7 | 1006 | 58 |
BC008853.2 | - | 6 - 7 | 782 | 58 | |
BC019016.2 | - | 6 - 7 | 801 | 58 | |
BI918365.1 | - | 5 - 6 | 498 | 58 | |
CF132216.1 | - | 5 - 6 | 523 | 58 |