The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144979.4 | NP_659416.1 | 4 - 5 | 1580 | 92 |
XM_011531692.1 | XP_011529994.1 | 4 - 5 | 1531 | 92 | |
XM_011531693.1 | XP_011529995.1 | 4 - 5 | 1583 | 92 | |
XM_011531694.1 | XP_011529996.1 | 4 - 5 | 1434 | 92 | |
XM_011531695.2 | XP_011529997.1 | 4 - 5 | 1479 | 92 | |
XM_011531696.1 | XP_011529998.1 | 4 - 5 | 1434 | 92 | |
GenBank mRNA | BC028588.2 | - | 4 - 5 | 1504 | 92 |