The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020546.2 | NP_065433.2 | 1 - 2 | 275 | 57 |
XM_011513942.2 | XP_011512244.1 | 1 - 2 | 222 | 57 | |
GenBank mRNA | AF410885.1 | - | 1 - 2 | 234 | 57 |
AK127036.1 | - | 1 - 2 | 223 | 57 | |
AK304041.1 | - | 1 - 2 | 110 | 57 | |
BC111743.1 | - | 1 - 2 | 213 | 57 | |
BE677757.1 | - | 1 - 2 | 273 | 57 | |
DB481853.2 | - | 1 - 2 | 275 | 57 |