The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020546.2 | NP_065433.2 | 21 - 22 | 2846 | 82 |
XM_011513942.2 | XP_011512244.1 | 20 - 21 | 2655 | 82 | |
GenBank mRNA | AB028983.1 | - | 18 - 19 | 2167 | 82 |
AF070583.1 | - | 5 - 6 | 607 | 82 | |
AF410885.1 | - | 21 - 22 | 2805 | 82 | |
AK123013.1 | - | 21 - 22 | 2693 | 82 | |
AK127036.1 | - | 21 - 22 | 2795 | 82 | |
AK294555.1 | - | 18 - 19 | 2302 | 82 | |
AK295531.1 | - | 18 - 19 | 2186 | 82 | |
AK299775.1 | - | 21 - 22 | 2272 | 82 | |
AK304041.1 | - | 21 - 22 | 2681 | 82 | |
AK307474.1 | - | 4 - 5 | 490 | 82 | |
BC111743.1 | - | 21 - 22 | 2784 | 82 | |
CR749634.1 | - | 1 - 2 | 8748 | 82 | |
L21993.1 | - | 8 - 9 | 934 | 82 | |
X74210.1 | - | 8 - 9 | 934 | 82 |