The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165979.2 | NP_001159451.1 | 6 - 7 | 1560 | 134 |
NM_001288989.1 | NP_001275918.1 | 7 - 8 | 3049 | 134 | |
NM_016341.3 | NP_057425.3 | 7 - 8 | 3049 | 134 | |
XM_006717885.3 | XP_006717948.1 | 7 - 8 | 2997 | 134 | |
XM_006717888.3 | XP_006717951.1 | 7 - 8 | 2997 | 134 | |
XM_006717889.3 | XP_006717952.1 | 7 - 8 | 2997 | 134 | |
XM_006717890.2 | XP_006717953.1 | 6 - 7 | 1513 | 134 | |
XM_011539849.2 | XP_011538151.1 | 7 - 8 | 4154 | 134 | |
XM_011539850.2 | XP_011538152.1 | 6 - 7 | 2642 | 134 | |
XM_011539851.2 | XP_011538153.1 | 7 - 8 | 2999 | 134 | |
XM_011539852.2 | XP_011538154.1 | 7 - 8 | 2999 | 134 | |
XM_017016310.1 | XP_016871799.1 | 8 - 9 | 3044 | 134 | |
XM_017016311.1 | XP_016871800.1 | 7 - 8 | 2997 | 134 | |
XM_017016312.1 | XP_016871801.1 | 6 - 7 | 1513 | 134 | |
GenBank mRNA | AB040949.2 | - | 7 - 8 | 3049 | 134 |
AF170071.1 | - | 6 - 7 | 1491 | 134 | |
AK289852.1 | - | 6 - 7 | 1513 | 134 | |
AK297779.1 | - | 6 - 7 | 1300 | 134 | |
BC140705.1 | - | 6 - 7 | 2475 | 134 | |
BC144286.1 | - | 6 - 7 | 2475 | 134 | |
BC151854.1 | - | 7 - 8 | 3049 | 134 |