The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152305.2 | NP_689518.1 | 7 - 8 | 820 | 109 |
NR_024265.1 | 7 - 8 | 963 | 109 | ||
XM_006713705.3 | XP_006713768.1 | 7 - 8 | 772 | 109 | |
XM_017006878.1 | XP_016862367.1 | 7 - 8 | 935 | 109 | |
XM_017006879.1 | XP_016862368.1 | 8 - 9 | 1081 | 109 | |
XM_017006880.1 | XP_016862369.1 | 8 - 9 | 938 | 109 | |
GenBank mRNA | AF168711.1 | - | 7 - 8 | 771 | 109 |
AK075444.1 | - | 7 - 8 | 756 | 109 | |
AK222937.1 | - | 7 - 8 | 780 | 109 | |
AK295984.1 | - | 7 - 8 | 938 | 109 | |
AK315367.1 | - | 7 - 8 | 795 | 109 | |
AY298903.1 | - | 7 - 8 | 806 | 109 | |
AY358581.1 | - | 7 - 8 | 763 | 109 | |
BC030614.2 | - | 7 - 8 | 806 | 109 | |
BC048810.1 | - | 7 - 8 | 764 | 109 |