The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014849.4 | NP_055664.3 | 6 - 7 | 1669 | 93 |
XM_017002992.1 | XP_016858481.1 | 6 - 7 | 1455 | 93 | |
GenBank mRNA | AB018279.1 | - | 6 - 7 | 1612 | 93 |
AK075480.1 | - | 6 - 7 | 1629 | 93 | |
AK291718.1 | - | 6 - 7 | 1618 | 93 | |
BC000776.2 | - | 5 - 6 | 764 | 93 | |
BC045111.1 | - | 6 - 7 | 1615 | 93 | |
BX537754.1 | - | 6 - 7 | 1665 | 93 |