The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185085.1 | NP_001172014.1 | 18 - 19 | 2964 | 72 |
NM_001676.5 | NP_001667.4 | 18 - 19 | 2946 | 72 | |
GenBank mRNA | AK292968.1 | - | 18 - 19 | 2946 | 72 |
AK303498.1 | - | 11 - 12 | 1698 | 72 | |
BC031609.1 | - | 18 - 19 | 2798 | 72 | |
U02076.1 | - | 18 - 19 | 2780 | 72 |