The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291571.1 | NP_001278500.1 | 17 - 18 | 2816 | 125 |
NM_005899.4 | NP_005890.2 | 17 - 18 | 2816 | 125 | |
NM_031862.3 | NP_114068.1 | 17 - 18 | 2624 | 125 | |
XM_006721903.1 | XP_006721966.1 | 16 - 17 | 2388 | 125 | |
XM_011524813.1 | XP_011523115.1 | 17 - 18 | 2925 | 125 | |
XM_017024642.1 | XP_016880131.1 | 18 - 19 | 3928 | 125 | |
XM_017024643.1 | XP_016880132.1 | 16 - 17 | 2715 | 125 | |
XM_017024644.1 | XP_016880133.1 | 16 - 17 | 2414 | 125 | |
GenBank mRNA | AK290005.1 | - | 17 - 18 | 2572 | 125 |
AK297425.1 | - | 7 - 8 | 684 | 125 | |
AY450308.1 | - | 17 - 18 | 2538 | 125 | |
BC009808.2 | - | 17 - 18 | 2543 | 125 | |
D30756.1 | - | 17 - 18 | 2614 | 125 | |
X76952.1 | - | 17 - 18 | 2566 | 125 |