The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291571.1 | NP_001278500.1 | 3 - 4 | 501 | 120 |
NM_005899.4 | NP_005890.2 | 3 - 4 | 501 | 120 | |
NM_031862.3 | NP_114068.1 | 3 - 4 | 309 | 120 | |
XM_006721903.1 | XP_006721966.1 | 3 - 4 | 283 | 120 | |
XM_011524813.1 | XP_011523115.1 | 3 - 4 | 610 | 120 | |
XM_017024642.1 | XP_016880131.1 | 4 - 5 | 1613 | 120 | |
XM_017024643.1 | XP_016880132.1 | 3 - 4 | 610 | 120 | |
XM_017024644.1 | XP_016880133.1 | 3 - 4 | 309 | 120 | |
GenBank mRNA | AK290005.1 | - | 3 - 4 | 257 | 120 |
AK297425.1 | - | 3 - 4 | 246 | 120 | |
AY450308.1 | - | 3 - 4 | 223 | 120 | |
BC009808.2 | - | 3 - 4 | 228 | 120 | |
D30756.1 | - | 3 - 4 | 299 | 120 | |
DB081342.1 | - | 3 - 4 | 309 | 120 | |
X76952.1 | - | 3 - 4 | 251 | 120 |