The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282116.1 | NP_001269045.1 | 4 - 5 | 736 | 88 |
NM_001282117.1 | NP_001269046.1 | 5 - 6 | 720 | 88 | |
NM_002919.3 | NP_002910.1 | 3 - 4 | 481 | 88 | |
NM_134428.2 | NP_602304.1 | 5 - 6 | 803 | 88 | |
XM_006716840.1 | XP_006716903.1 | 4 - 5 | 649 | 88 | |
XM_006716843.1 | XP_006716906.1 | 5 - 6 | 649 | 88 | |
XM_006716844.1 | XP_006716907.1 | 5 - 6 | 665 | 88 | |
XM_006716845.2 | XP_006716908.1 | 4 - 5 | 727 | 88 | |
XM_011517998.1 | XP_011516300.1 | 5 - 6 | 717 | 88 | |
XM_011517999.1 | XP_011516301.1 | 5 - 6 | 681 | 88 | |
XM_011518001.1 | XP_011516303.1 | 6 - 7 | 731 | 88 | |
XM_017015000.1 | XP_016870489.1 | 5 - 6 | 668 | 88 | |
GenBank mRNA | AK289540.1 | - | 4 - 5 | 534 | 88 |
BC022191.2 | - | 5 - 6 | 784 | 88 | |
BC067778.1 | - | 5 - 6 | 662 | 88 | |
X76092.1 | - | 3 - 4 | 481 | 88 |