Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPECC1
Entrez Gene ID
92521
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
Gene Aliases
CYTSB, HCMOGT-1, HCMOGT1, NSP
Chromosome Location
Chr.17: 20009301 - 20319026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033554.2 NP_001028726.1 5 - 6 2312 105
NM_152904.4 NP_690868.3 7 - 8 2403 105
XM_017025322.1 XP_016880811.1 6 - 7 2379 105
XM_017025325.1 XP_016880814.1 7 - 8 2497 105
GenBank mRNA AB041533.1 - 7 - 8 2403 105
AY816326.1 - 5 - 6 2217 105
AY816328.1 - 7 - 8 2403 105
BC033618.1 - 4 - 5 1427 105

View Full Product Details