Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP98
Entrez Gene ID
4928
Gene Name
nucleoporin 98
Gene Aliases
ADIR2, NUP196, NUP96
Chromosome Location
Chr.11: 3675010 - 3797792 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016320.4 NP_057404.2 22 - 23 3500 81
NM_139132.3 NP_624358.2 22 - 23 3500 81
GenBank mRNA AF071076.1 - 22 - 23 3203 81
AF071077.1 - 22 - 23 3203 81
AF231130.1 - 22 - 23 3223 81
BC012906.1 - 2 - 3 245 81

View Full Product Details