Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP98
Entrez Gene ID
4928
Gene Name
nucleoporin 98
Gene Aliases
ADIR2, NUP196, NUP96
Chromosome Location
Chr.11: 3675010 - 3797792 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016320.4 NP_057404.2 24 - 25 4169 77
NM_139132.3 NP_624358.2 24 - 25 4169 77
GenBank mRNA AF071076.1 - 24 - 25 3872 77
AF071077.1 - 24 - 25 3872 77
AF116074.1 - 2 - 3 276 77
AF231130.1 - 24 - 25 3892 77
AL133601.1 - 1 - 2 123 77
BC012906.1 - 4 - 5 605 77
BT007349.1 - 1 - 2 166 77

View Full Product Details