Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUP98
Entrez Gene ID
4928
Gene Name
nucleoporin 98
Gene Aliases
ADIR2, NUP196, NUP96
Chromosome Location
Chr.11: 3675010 - 3797792 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016320.4 NP_057404.2 32 - 33 5600 78
NM_139132.3 NP_624358.2 31 - 32 5378 78
GenBank mRNA AF071076.1 - 32 - 33 5323 78
AF071077.1 - 31 - 32 5101 78
AF116074.1 - 10 - 11 1706 78
AF231130.1 - 32 - 33 5323 78
AL133601.1 - 9 - 10 1554 78
AL137613.1 - 4 - 5 674 78
BC012906.1 - 12 - 13 2036 78
BT007349.1 - 9 - 10 1597 78

View Full Product Details