Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LVRN
Entrez Gene ID
206338
Gene Name
laeverin
Gene Aliases
APQ, AQPEP, TAQPEP
Chromosome Location
Chr.5: 115962440 - 116027602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173800.4 NP_776161.3 4 - 5 1271 111
XM_011543263.2 XP_011541565.1 4 - 5 1285 111
XM_011543264.2 XP_011541566.1 4 - 5 1285 111
XM_017009215.1 XP_016864704.1 4 - 5 1285 111
GenBank mRNA AK291655.1 - 4 - 5 1285 111
AY560010.1 - 4 - 5 1159 111
BC060869.1 - 5 - 6 1198 111
BC068560.1 - 4 - 5 1271 111
BC109022.1 - 4 - 5 1135 111
BC109023.1 - 4 - 5 1135 111

View Full Product Details