Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA2D4
Entrez Gene ID
93589
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Gene Aliases
RCD4
Chromosome Location
Chr.12: 1791957 - 1922606 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172364.4 NP_758952.4 31 - 32 3102 70
XM_011521041.2 XP_011519343.1 30 - 31 6941 70
GenBank mRNA AF516695.1 - 31 - 32 3234 70
AK301234.1 - 4 - 5 343 70
AL137658.1 - 8 - 9 641 70
BC048288.1 - 3 - 4 208 70
BC150186.1 - 30 - 31 3025 70
BX537436.1 - 31 - 32 3065 70
BX537437.1 - 31 - 32 3103 70

View Full Product Details