The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016148.3 | NP_057232.2 | 10 - 11 | 1569 | 147 |
XM_006723233.3 | XP_006723296.1 | 11 - 12 | 1967 | 147 | |
XM_011527013.2 | XP_011525315.1 | 11 - 12 | 1967 | 147 | |
XM_011527014.2 | XP_011525316.1 | 11 - 12 | 1967 | 147 | |
GenBank mRNA | AF163302.1 | - | 10 - 11 | 1569 | 147 |