The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000030.2 | NP_000021.1 | 6 - 7 | 795 | 146 |
GenBank mRNA | AF348451.1 | - | 6 - 7 | 146 | |
AK292754.1 | - | 6 - 7 | 716 | 146 | |
AK315331.1 | - | 6 - 7 | 743 | 146 | |
BC132819.1 | - | 6 - 7 | 792 | 146 | |
D13368.1 | - | 6 - 7 | 782 | 146 | |
X53414.1 | - | 6 - 7 | 796 | 146 | |
X56092.1 | - | 6 - 7 | 695 | 146 |