The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171805.1 | NP_001165276.1 | 10 - 11 | 1231 | 58 |
NM_001171806.1 | NP_001165277.1 | 4 - 5 | 833 | 58 | |
NM_006999.4 | NP_008930.1 | 10 - 11 | 1231 | 58 | |
XM_005248234.3 | XP_005248291.1 | 10 - 11 | 1884 | 58 | |
XM_011513945.1 | XP_011512247.1 | 8 - 9 | 1007 | 58 | |
XM_017008988.1 | XP_016864477.1 | 10 - 11 | 2505 | 58 | |
XM_017008989.1 | XP_016864478.1 | 10 - 11 | 1143 | 58 | |
XM_017008990.1 | XP_016864479.1 | 5 - 6 | 1689 | 58 | |
GenBank mRNA | AB005754.3 | - | 9 - 10 | 1139 | 58 |
AF089896.1 | - | 10 - 11 | 1223 | 58 | |
AK289857.1 | - | 9 - 10 | 1233 | 58 | |
AK303516.1 | - | 4 - 5 | 789 | 58 | |
AK308405.1 | - | 4 - 5 | 833 | 58 | |
BC084567.1 | - | 10 - 11 | 1164 | 58 | |
BC117137.1 | - | 9 - 10 | 1149 | 58 | |
BC126106.1 | - | 9 - 10 | 1149 | 58 | |
BC143882.1 | - | 9 - 10 | 1149 | 58 | |
KC424495.1 | - | 10 - 11 | 1792 | 58 |