Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM7
Entrez Gene ID
81786
Gene Name
tripartite motif containing 7
Gene Aliases
GNIP, RNF90
Chromosome Location
Chr.5: 181193924 - 181206000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203296.1 NP_976041.1 1 - 2 596 91
XM_017009903.1 XP_016865392.1 1 - 2 676 91
GenBank mRNA AF396654.1 - 1 - 2 380 91

View Full Product Details