The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100603.1 | NP_001094073.1 | 2 - 3 | 349 | 104 |
NM_006854.3 | NP_006845.1 | 2 - 3 | 349 | 104 | |
GenBank mRNA | AK315702.1 | - | 2 - 3 | 321 | 104 |
AL547016.3 | - | 2 - 3 | 326 | 104 | |
BC008081.2 | - | 2 - 3 | 253 | 104 | |
BC012994.2 | - | 2 - 3 | 284 | 104 | |
BC071982.1 | - | 2 - 3 | 303 | 104 | |
DA713413.1 | - | 2 - 3 | 349 | 104 | |
M88458.1 | - | 2 - 3 | 226 | 104 | |
X63745.1 | - | 2 - 3 | 200 | 104 |