Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAIP
Entrez Gene ID
10293
Gene Name
TRAF interacting protein
Gene Aliases
RNF206, SCKL9, TRIP
Chromosome Location
Chr.3: 49828595 - 49856584 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005879.2 NP_005870.2 10 - 11 987 78
XM_011533264.1 XP_011531566.1 10 - 11 1081 78
XM_017005526.1 XP_016861015.1 7 - 8 715 78
GenBank mRNA AB451320.1 - 10 - 11 881 78
AB451449.1 - 10 - 11 881 78
AK225873.1 - 7 - 8 666 78
AK292172.1 - 10 - 11 975 78
BC000310.2 - 10 - 11 968 78
BC019283.1 - 10 - 11 968 78
U77845.1 - 10 - 11 983 78

View Full Product Details