The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031862.3 | NP_114068.1 | 1 - 2 | 146 | 113 |
XM_017024644.1 | XP_016880133.1 | 1 - 2 | 146 | 113 | |
GenBank mRNA | D30756.1 | - | 1 - 2 | 136 | 113 |
DB081342.1 | - | 1 - 2 | 146 | 113 |