Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED1
Entrez Gene ID
5469
Gene Name
mediator complex subunit 1
Gene Aliases
CRSP1, CRSP200, DRIP205, DRIP230, PBP, PPARBP, PPARGBP, RB18A, TRAP220, TRIP2
Chromosome Location
Chr.17: 39404285 - 39451281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004774.3 NP_004765.2 15 - 16 1605 101
XM_005257465.3 XP_005257522.1 14 - 15 1479 101
XM_006721957.1 XP_006722020.1 15 - 16 1602 101
XM_017024779.1 XP_016880268.1 14 - 15 1538 101
GenBank mRNA AF055994.1 - 15 - 16 1585 101
AF283812.1 - 14 - 15 1348 101
AK299789.1 - 14 - 15 1470 101
AK314185.1 - 15 - 16 1602 101
BC006517.2 - 15 - 16 1578 101
BC060758.1 - 15 - 16 1583 101
BC131783.1 - 15 - 16 1459 101
Y13467.1 - 15 - 16 1583 101

View Full Product Details