Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFI1B
Entrez Gene ID
8328
Gene Name
growth factor independent 1B transcriptional repressor
Gene Aliases
BDPLT17, ZNF163B
Chromosome Location
Chr.9: 132945545 - 132991697 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006717297.3 XP_006717360.1 1 - 2 296 83

View Full Product Details