The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320509.1 | NP_001307438.1 | 4 - 5 | 657 | 60 |
XM_017024457.1 | XP_016879946.1 | 4 - 5 | 458 | 60 |