The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316983.1 | NP_001303912.1 | 5 - 6 | 630 | 53 |
NM_001316984.1 | NP_001303913.1 | 5 - 6 | 588 | 53 | |
NM_001316985.1 | NP_001303914.1 | 5 - 6 | 585 | 53 | |
NM_001316988.1 | NP_001303917.1 | 5 - 6 | 568 | 53 | |
NM_058190.3 | NP_478070.1 | 5 - 6 | 633 | 53 | |
XM_017028481.1 | XP_016883970.1 | 5 - 6 | 781 | 53 | |
XM_017028482.1 | XP_016883971.1 | 4 - 5 | 521 | 53 | |
GenBank mRNA | AF391113.1 | - | 5 - 6 | 614 | 53 |
AF391114.1 | - | 5 - 6 | 569 | 53 | |
BC009341.2 | - | 5 - 6 | 590 | 53 | |
BG744869.1 | - | 1 - 2 | 53 | ||
BM042090.1 | - | 5 - 6 | 572 | 53 | |
CN368049.1 | - | 5 - 6 | 618 | 53 |