The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127190.1 | NP_001120662.1 | 6 - 7 | 642 | 78 |
NM_004383.2 | NP_004374.1 | 5 - 6 | 1161 | 78 | |
XM_005254165.4 | XP_005254222.1 | 5 - 6 | 611 | 78 | |
XM_017021925.1 | XP_016877414.1 | 5 - 6 | 1202 | 78 | |
GenBank mRNA | AB451288.1 | - | 4 - 5 | 459 | 78 |
AB451420.1 | - | 4 - 5 | 459 | 78 | |
AK127672.1 | - | 7 - 8 | 891 | 78 | |
AK223626.1 | - | 5 - 6 | 722 | 78 | |
AK290531.1 | - | 6 - 7 | 633 | 78 | |
AK297694.1 | - | 3 - 4 | 435 | 78 | |
AK298245.1 | - | 5 - 6 | 577 | 78 | |
AK309344.1 | - | 1 - 2 | 174 | 78 | |
AK312669.1 | - | 5 - 6 | 664 | 78 | |
AY007162.1 | - | 3 - 4 | 391 | 78 | |
BC104847.1 | - | 5 - 6 | 619 | 78 | |
BC104875.1 | - | 5 - 6 | 619 | 78 | |
BC106073.1 | - | 5 - 6 | 1161 | 78 | |
CR541960.1 | - | 4 - 5 | 459 | 78 | |
X59932.1 | - | 5 - 6 | 592 | 78 | |
X60114.1 | - | 5 - 6 | 871 | 78 |