The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127190.1 | NP_001120662.1 | 7 - 8 | 737 | 59 |
NM_004383.2 | NP_004374.1 | 6 - 7 | 1256 | 59 | |
XM_005254165.4 | XP_005254222.1 | 6 - 7 | 706 | 59 | |
XM_017021925.1 | XP_016877414.1 | 6 - 7 | 1297 | 59 | |
GenBank mRNA | AB451288.1 | - | 5 - 6 | 554 | 59 |
AB451420.1 | - | 5 - 6 | 554 | 59 | |
AK127672.1 | - | 8 - 9 | 986 | 59 | |
AK223626.1 | - | 6 - 7 | 817 | 59 | |
AK297694.1 | - | 4 - 5 | 530 | 59 | |
AK298245.1 | - | 6 - 7 | 672 | 59 | |
AK312669.1 | - | 6 - 7 | 759 | 59 | |
AY007162.1 | - | 4 - 5 | 486 | 59 | |
BC104847.1 | - | 6 - 7 | 714 | 59 | |
BC104875.1 | - | 6 - 7 | 714 | 59 | |
BC106073.1 | - | 6 - 7 | 1256 | 59 | |
CR541960.1 | - | 5 - 6 | 554 | 59 | |
X59932.1 | - | 6 - 7 | 687 | 59 | |
X60114.1 | - | 6 - 7 | 966 | 59 |