The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207014.1 | NP_001193943.1 | 1 - 2 | 261 | 53 |
GenBank mRNA | AK290124.1 | - | 1 - 2 | 198 | 53 |
AK309905.1 | - | 1 - 2 | 64 | 53 | |
DC398178.1 | - | 1 - 2 | 262 | 53 |