The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085454.1 | NP_001078923.1 | 1 - 2 | 271 | 67 |
NM_014030.3 | NP_054749.2 | 1 - 2 | 271 | 67 | |
XM_011524684.1 | XP_011522986.1 | 1 - 2 | 112 | 67 | |
XM_011524685.1 | XP_011522987.1 | 1 - 2 | 112 | 67 | |
GenBank mRNA | AB209537.1 | - | 1 - 2 | 70 | 67 |
AF124490.1 | - | 1 - 2 | 71 | 67 | |
AK294785.1 | - | 2 - 3 | 149 | 67 | |
AK299932.1 | - | 1 - 2 | 115 | 67 | |
BC048196.1 | - | 1 - 2 | 298 | 67 | |
BC067358.1 | - | 1 - 2 | 271 | 67 | |
CF994106.1 | - | 1 - 2 | 333 | 67 |