The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142556.1 | NP_001136028.1 | 8 - 9 | 910 | 147 |
NM_001142557.1 | NP_001136029.1 | 5 - 6 | 683 | 147 | |
NM_012484.2 | NP_036616.2 | 8 - 9 | 907 | 147 | |
NM_012485.2 | NP_036617.2 | 7 - 8 | 862 | 147 | |
GenBank mRNA | AF032862.1 | - | 8 - 9 | 761 | 147 |
AK290577.1 | - | 7 - 8 | 703 | 147 | |
AK302975.1 | - | 9 - 10 | 717 | 147 | |
AK303616.1 | - | 5 - 6 | 561 | 147 | |
AK313603.1 | - | 8 - 9 | 828 | 147 | |
AY623426.1 | - | 8 - 9 | 733 | 147 | |
BC017793.1 | - | 7 - 8 | 731 | 147 | |
BC030505.2 | - | 8 - 9 | 817 | 147 | |
BC057235.1 | - | 7 - 8 | 832 | 147 | |
BC070281.1 | - | 7 - 8 | 738 | 147 | |
BC108904.1 | - | 8 - 9 | 762 | 147 | |
U29343.1 | - | 8 - 9 | 788 | 147 |