Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B9D1
Entrez Gene ID
27077
Gene Name
B9 domain containing 1
Gene Aliases
B9, EPPB9, MKS9, MKSR1
Chromosome Location
Chr.17: 19335372 - 19378193 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015681.4 NP_056496.1 6 - 7 61
GenBank mRNA AB030506.1 - 6 - 7 61
AI300844.1 - 1 - 2 298 61

View Full Product Details