The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020197.2 | NP_064582.2 | 2 - 3 | 263 | 75 |
GenBank mRNA | AF226053.1 | - | 2 - 3 | 252 | 75 |
AK313868.1 | - | 2 - 3 | 233 | 75 | |
BC098133.1 | - | 2 - 3 | 252 | 75 | |
BC098276.1 | - | 2 - 3 | 252 | 75 | |
BC098305.1 | - | 2 - 3 | 252 | 75 | |
BC098335.1 | - | 2 - 3 | 252 | 75 |