The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098511.2 | NP_001091981.1 | 7 - 8 | 957 | 138 |
NM_001243952.1 | NP_001230881.1 | 8 - 9 | 1160 | 138 | |
NM_001243953.1 | NP_001230882.1 | 7 - 8 | 900 | 138 | |
NM_004520.4 | NP_004511.2 | 7 - 8 | 957 | 138 | |
GenBank mRNA | AK297347.1 | - | 7 - 8 | 703 | 138 |
AK302270.1 | - | 7 - 8 | 594 | 138 | |
AK307880.1 | - | 7 - 8 | 611 | 138 | |
AK315866.1 | - | 7 - 8 | 584 | 138 | |
AY317140.1 | - | 5 - 6 | 488 | 138 | |
BC031828.2 | - | 6 - 7 | 582 | 138 | |
EF560716.1 | - | 7 - 8 | 668 | 138 | |
EF560728.1 | - | 7 - 8 | 662 | 138 | |
Y08319.1 | - | 6 - 7 | 583 | 138 |