The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024830.3 | NP_079106.3 | 9 - 10 | 1020 | 117 |
XM_005248373.3 | XP_005248430.1 | 10 - 11 | 928 | 117 | |
XM_011514132.1 | XP_011512434.1 | 9 - 10 | 1029 | 117 | |
XM_011514134.1 | XP_011512436.1 | 9 - 10 | 795 | 117 | |
XM_017009879.1 | XP_016865368.1 | 3 - 4 | 331 | 117 | |
GenBank mRNA | AB244719.2 | - | 9 - 10 | 903 | 117 |
AK022499.1 | - | 4 - 5 | 199 | 117 | |
AK022505.1 | - | 4 - 5 | 199 | 117 | |
AK090444.1 | - | 7 - 8 | 499 | 117 | |
BC020166.1 | - | 7 - 8 | 486 | 117 |