The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122670.1 | NP_001116142.1 | 11 - 12 | 1441 | 66 |
NM_001172436.1 | NP_001165907.1 | 11 - 12 | 1441 | 66 | |
NM_002637.3 | NP_002628.2 | 11 - 12 | 1441 | 66 | |
XM_006724661.2 | XP_006724724.1 | 11 - 12 | 1283 | 66 | |
GenBank mRNA | BC104944.1 | - | 11 - 12 | 1187 | 66 |
BC143499.1 | - | 11 - 12 | 1187 | 66 | |
BC143501.1 | - | 11 - 12 | 1187 | 66 | |
X73874.1 | - | 11 - 12 | 1301 | 66 |