The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122670.1 | NP_001116142.1 | 13 - 14 | 1621 | 117 |
NM_001172436.1 | NP_001165907.1 | 13 - 14 | 1621 | 117 | |
NM_002637.3 | NP_002628.2 | 13 - 14 | 1621 | 117 | |
XM_006724661.2 | XP_006724724.1 | 13 - 14 | 1463 | 117 | |
GenBank mRNA | BC104944.1 | - | 13 - 14 | 1367 | 117 |
BC143499.1 | - | 13 - 14 | 1367 | 117 | |
BC143501.1 | - | 13 - 14 | 1367 | 117 | |
X73874.1 | - | 13 - 14 | 1481 | 117 |