The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122670.1 | NP_001116142.1 | 19 - 20 | 2431 | 77 |
NM_002637.3 | NP_002628.2 | 19 - 20 | 2431 | 77 | |
GenBank mRNA | BC104944.1 | - | 19 - 20 | 2177 | 77 |
BC143499.1 | - | 19 - 20 | 2177 | 77 | |
X73874.1 | - | 19 - 20 | 2291 | 77 |