Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGM4
Entrez Gene ID
7047
Gene Name
transglutaminase 4
Gene Aliases
TGP, hTGP
Chromosome Location
Chr.3: 44874592 - 44914601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003241.3 NP_003232.2 6 - 7 723 84
XM_011534042.1 XP_011532344.1 7 - 8 872 84
GenBank mRNA AK300523.1 - 6 - 7 592 84
AK312936.1 - 6 - 7 721 84
BC007003.1 - 6 - 7 718 84
L34840.1 - 6 - 7 694 84
U31905.1 - 6 - 7 694 84
U79008.1 - 5 - 6 587 84
Y09559.1 - 5 - 6 587 84

View Full Product Details