The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167827.2 | NP_001161299.2 | 3 - 4 | 610 | 61 |
NM_001271944.1 | NP_001258873.1 | 3 - 4 | 674 | 61 | |
NM_001271945.1 | NP_001258874.1 | 3 - 4 | 585 | 61 | |
NM_007162.2 | NP_009093.1 | 4 - 5 | 776 | 61 | |
XM_005249411.1 | XP_005249468.1 | 3 - 4 | 709 | 61 | |
XM_005249412.1 | XP_005249469.1 | 3 - 4 | 601 | 61 | |
XM_006715212.3 | XP_006715275.1 | 3 - 4 | 828 | 61 | |
XM_006715213.2 | XP_006715276.1 | 3 - 4 | 652 | 61 | |
XM_011514915.1 | XP_011513217.1 | 3 - 4 | 831 | 61 | |
XM_011514916.2 | XP_011513218.1 | 4 - 5 | 621 | 61 | |
GenBank mRNA | AJ608786.1 | - | 3 - 4 | 610 | 61 |
AK095061.1 | - | 3 - 4 | 1701 | 61 | |
AK298390.1 | - | 3 - 4 | 595 | 61 | |
AK310063.1 | - | 3 - 4 | 832 | 61 | |
BC032448.1 | - | 4 - 5 | 776 | 61 |