The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000442.4 | NP_000433.4 | 12 - 13 | 2259 | 124 |
XM_005276880.1 | XP_005276937.1 | 12 - 13 | 2545 | 124 | |
XM_005276881.1 | XP_005276938.1 | 12 - 13 | 2545 | 124 | |
XM_011524889.2 | XP_011523191.1 | 13 - 14 | 2275 | 124 | |
XM_011524890.1 | XP_011523192.1 | 13 - 14 | 2224 | 124 | |
XM_017024738.1 | XP_016880227.1 | 13 - 14 | 2168 | 124 | |
XM_017024740.1 | XP_016880229.1 | 13 - 14 | 2238 | 124 | |
GenBank mRNA | AF281287.1 | - | 12 - 13 | 2129 | 124 |
AF281288.1 | - | 12 - 13 | 2129 | 124 | |
AF281289.1 | - | 12 - 13 | 2129 | 124 | |
AF281290.1 | - | 12 - 13 | 2129 | 124 | |
AF281291.1 | - | 12 - 13 | 2129 | 124 | |
AF281292.1 | - | 12 - 13 | 2129 | 124 | |
AF281293.1 | - | 12 - 13 | 2129 | 124 | |
AF281294.1 | - | 12 - 13 | 2129 | 124 | |
AF281295.1 | - | 12 - 13 | 2129 | 124 | |
AF281296.1 | - | 12 - 13 | 2129 | 124 | |
AF281297.1 | - | 12 - 13 | 2129 | 124 | |
AF281298.1 | - | 12 - 13 | 2129 | 124 | |
AF281299.1 | - | 12 - 13 | 2129 | 124 | |
AF281300.1 | - | 12 - 13 | 2129 | 124 | |
AF281301.1 | - | 12 - 13 | 2129 | 124 | |
AF393676.1 | - | 12 - 13 | 2129 | 124 | |
AF393677.1 | - | 12 - 13 | 2129 | 124 | |
AF393678.1 | - | 12 - 13 | 2129 | 124 | |
AK290692.1 | - | 12 - 13 | 2062 | 124 | |
AK303959.1 | - | 7 - 8 | 1033 | 124 | |
AK304166.1 | - | 11 - 12 | 2059 | 124 | |
BC022512.1 | - | 12 - 13 | 2239 | 124 | |
BC051822.1 | - | 12 - 13 | 2259 | 124 | |
JQ287500.1 | - | 12 - 13 | 2046 | 124 | |
M28526.1 | - | 12 - 13 | 2187 | 124 | |
M37780.1 | - | 12 - 13 | 2171 | 124 |