The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320910.1 | NP_001307839.1 | 7 - 8 | 1201 | 79 |
NM_002041.4 | NP_002032.2 | 6 - 7 | 1156 | 79 | |
NM_005254.5 | NP_005245.2 | 6 - 7 | 1156 | 79 | |
NM_016654.4 | NP_057738.1 | 6 - 7 | 1120 | 79 | |
NM_016655.4 | NP_057739.1 | 6 - 7 | 1120 | 79 | |
NM_181427.3 | NP_852092.1 | 7 - 8 | 1144 | 79 | |
XM_005254274.3 | XP_005254331.1 | 6 - 7 | 864 | 79 | |
XM_017022053.1 | XP_016877542.1 | 7 - 8 | 899 | 79 | |
GenBank mRNA | AK303901.1 | - | 5 - 6 | 764 | 79 |
BC016910.1 | - | 6 - 7 | 841 | 79 | |
BC036080.1 | - | 6 - 7 | 914 | 79 | |
BC050702.1 | - | 6 - 7 | 861 | 79 | |
BQ573816.1 | - | 2 - 3 | 80 | ||
BT006652.1 | - | 5 - 6 | 732 | 79 | |
D13317.1 | - | 6 - 7 | 901 | 79 | |
EU159454.1 | - | 5 - 6 | 734 | 79 | |
U13045.1 | - | 6 - 7 | 901 | 79 | |
U13046.1 | - | 6 - 7 | 865 | 79 | |
U13047.1 | - | 6 - 7 | 901 | 79 | |
U13048.1 | - | 6 - 7 | 865 | 79 |