The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005334.2 | NP_005325.2 | 25 - 26 | 6592 | 75 |
XM_006724815.2 | XP_006724878.1 | 25 - 26 | 7192 | 75 | |
XM_006724816.2 | XP_006724879.1 | 25 - 26 | 7189 | 75 | |
XM_011531147.2 | XP_011529449.1 | 25 - 26 | 7069 | 75 | |
XM_011531148.2 | XP_011529450.1 | 25 - 26 | 7066 | 75 | |
XM_017029471.1 | XP_016884960.1 | 24 - 25 | 6991 | 75 | |
XM_017029472.1 | XP_016884961.1 | 20 - 21 | 5377 | 75 | |
GenBank mRNA | L20010.1 | - | 25 - 26 | 6408 | 75 |